Set of diaphragms and valves, for MD 4 NT, MV 2 NT, MD 4 NT VARIO, MV 2 NT VARIO 1 * 1 items
Catalog # VACU20696861
Supplier: VACUUBRAND
Set of diaphragms and valves, for MD 4 NT, MV 2 NT, MD 4 NT VARIO, MV 2 NT VARIO 1 * 1 items
Catalog # VACU20696861
Supplier: VACUUBRAND
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Grade 113V qualitative filter papers, fluted, Whatman™
Supplier: Whatman products (Cytiva)
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Mini vortex mixer
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Accessories for diaphragm pump
Supplier: VACUUBRAND