ContactUsLinkComponent
Rack, flipper 1, colour blue, material PP, autoclavable, temperature resistant -90°C to +120°C, dimension 197x95x115mm, 8 holes diam eter 30mm, 20 holes diameter 20mm. 1 * 1 items
Catalog # KOEH2-2773
Rack, flipper 1, colour blue, material PP, autoclavable, temperature resistant -90°C to +120°C, dimension 197x95x115mm, 8 holes diam eter 30mm, 20 holes diameter 20mm. 1 * 1 items
Catalog # KOEH2-2773
Frequently Bought Together