Orders LinkContactUsLinkComponent
Interfaces
Interfaces
Com.G@te®, external for Unistats®, CC®-E to CC®-205B
Catalog # HUBE6915
Supplier:   HUBER
Interfaces
Catalog # HUBE6915
Supplier:  HUBER
Interfaces
Interfaces
Com.G@te®, external for Unistats®, CC®-E to CC®-205B
Price
17 880,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Com.G@te®. The interfaces are integrated RS232 (bi-directional), RS485 (bi-directional), ECS external control signal, volt free contact, AIF analogue interface 0/4 to 20 mA or 0 to 10 V (bi-directional). Connections comply with NAMUR standard.
Specifications
  • Description:
    Com.G@te®, external for Unistats®, CC®-E to CC®-205B
  • Seq:
    02