ContactUsLinkComponent
DEN MCF STDS Set of liquid McFarland standards in 16mm ø glass tubes (0.5/1.0/2.0/3.0/4.0). Can be decanted into an alternative tube. Requires D16 adaptor. 1 * 5 items
Catalog # GRANDENMCFSTDS
Supplier:  GRANT INSTRUMENTS
undefined
DEN MCF STDS Set of liquid McFarland standards in 16mm ø glass tubes (0.5/1.0/2.0/3.0/4.0). Can be decanted into an alternative tube. Requires D16 adaptor. 1 * 5 items
Catalog # GRANDENMCFSTDS
Supplier:  GRANT INSTRUMENTS