Tubing, rubber, 8.0x2.5mm green. 1 * 10 m
Catalog # DENE91270825
Supplier: DEUTSCH NEUMANN
Frequently Bought Together
Petroleum spirit, 60…95 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Safety Pipette Fillers, Standard/Universal/Flip
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Disposable scalpels, sterile
Supplier: SWANN MORTON
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Stoppers, rubber, with reversible edge
Supplier: Saint Gobain Life Sciences
Tubing, rubber, 6.0x4.5mm white. 1 * 10 m
Supplier: DEUTSCH NEUMANN