ContactUsLinkComponent
Fixed Angle Rotor for 6 x 250ml Tubes, Max Speed: Ventilated 8,500rpm /11,228g, Refrigerated 9,500rpm/14,025g, With Aerosol-Tight lid,Rotor Angle: 25°, Autoclavable to 136°C. 1 * 1 items
Catalog # HETT5645
Supplier:  Hettich
Fixed Angle Rotor for 6 x 250ml Tubes, Max Speed: Ventilated 8,500rpm /11,228g, Refrigerated 9,500rpm/14,025g, With Aerosol-Tight lid,Rotor Angle: 25°, Autoclavable to 136°C. 1 * 1 items
Catalog # HETT5645
Supplier:  Hettich
Frequently Bought Together