ContactUsLinkComponent
Chemical Process Indicator Label for monitoring Ethylene Oxide (EO) sterilization processes. Labels are 1 x2 with Class 1 ISO 11 140-1:2005 compliant indicating strip. Labels are laser printable and provide visual assurance that a product has bee 1 * 1 Roll
Catalog # NAMSSP-E04
Supplier:  CROSSTEX
Chemical Process Indicator Label for monitoring Ethylene Oxide (EO) sterilization processes. Labels are 1 x2 with Class 1 ISO 11 140-1:2005 compliant indicating strip. Labels are laser printable and provide visual assurance that a product has bee 1 * 1 Roll
Catalog # NAMSSP-E04
Supplier:  CROSSTEX
Frequently Bought Together