2-ETHYLHEXYL 2-CYANO-3,3-DIPHENYL-ACRYLA TE, 97% 1 * 50 mL
Catalog # SIAL415820-50ML
Supplier: Merck
2-ETHYLHEXYL 2-CYANO-3,3-DIPHENYL-ACRYLA TE, 97% 1 * 50 mL
Catalog # SIAL415820-50ML
Supplier: Merck
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
DIMETHYLSULFOXIDE PURE 1 * 5 L
Supplier: Lach-ner S.R.O.
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
METHANOL SUITABLE FOR HPLC >99.9% 1 * 2,5 L
Supplier: Merck
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific