ContactUsLinkComponent
MINIMUM ESSENTIAL MEDIUM EAGLE, WITH EARLE'S SALTS AND SODIUM BICARBONATE, WITHOUT L-GLUTAMINE. STERILE-FILTERED. CELL 1 * 1 Each
Catalog # SIALM2279-24X500ML
Supplier:  Merck
MINIMUM ESSENTIAL MEDIUM EAGLE, WITH EARLE'S SALTS AND SODIUM BICARBONATE, WITHOUT L-GLUTAMINE. STERILE-FILTERED. CELL 1 * 1 Each
Catalog # SIALM2279-24X500ML
Supplier:  Merck
Frequently Bought Together