ContactUsLinkComponent
Box 10 x 10, green, 140 x 153 x 53 mm, compartment size 12.5 mm x 12.5 mm, made of polypropylene, autoclavable at 121°c and use in f reezer down to -90°c 1 * 1 items
Catalog # 479-1525
Supplier:  GLW
undefined
Box 10 x 10, green, 140 x 153 x 53 mm, compartment size 12.5 mm x 12.5 mm, made of polypropylene, autoclavable at 121°c and use in f reezer down to -90°c 1 * 1 items
Catalog # 479-1525
Supplier:  GLW
Frequently Bought Together