ContactUsLinkComponent
Box 10 x 10, blue, 140 x 153 x 53 mm, compartment size 12.5 mm x 12.5 mm, made of polypropylene, autoclavable at 121c and use in fr eezer down to -90c 1 * 1 items
Catalog # 479-1524
Supplier:  GLW
undefined
Box 10 x 10, blue, 140 x 153 x 53 mm, compartment size 12.5 mm x 12.5 mm, made of polypropylene, autoclavable at 121c and use in fr eezer down to -90c 1 * 1 items
Catalog # 479-1524
Supplier:  GLW
Frequently Bought Together