Areometer, DIN 12791, M50, 1,00-1,05:0,0010g/cm , ref. temp, 20°C, without thermometer max. 270mm long, suitable for government ver ification 1 * 1 items
Areometer, DIN 12791, M50, 1,00-1,05:0,0010g/cm , ref. temp, 20°C, without thermometer max. 270mm long, suitable for government ver ification 1 * 1 items