0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
PSEUDOMONAS ISOLATION AGAR, FOR MICRO-BI OLOGY 1 * 500 g
SIAL17208-500G
:
Merck
PSEUDOMONAS ISOLATION AGAR, FOR MICRO-BI OLOGY 1 * 500 g
SIAL17208-500G
:
Merck
Frequently Bought Together
BLOOD AGAR SHEEP 7% 90 MM PLATE 1 * 20 items
: LIOFILCHEM
VWR® Culture Media, Ready To Use, In Plates
: VWR Chemicals
Culture media, ready to use, Compact Dry
: Shimadzu Diagnostics Europe
Media supplements for dehydrated culture media
: THERMO FISHER DIAGNOSTICS
BPW (EP) + 0,3% TWEEN 80 1 * 6 items
: VWR Chemicals
V-MONOVETTE URINE 10,0 ML 100/15 MM ST 1 * 50 items
: SARSTEDT