POTASSIUM L-LACTATE SOLUTION, FOR MICROB IOLOGY 1 * 250 mL
Catalog # SIAL60389-250ML-F
Supplier: Merck
POTASSIUM L-LACTATE SOLUTION, FOR MICROB IOLOGY 1 * 250 mL
Catalog # SIAL60389-250ML-F
Supplier: Merck
Frequently Bought Together
Ethyl acetate ≥99.8%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
1-Butanol ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Methanol ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dichloromethane, anhydrous (max. 0.001% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals
Methanol, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific