(S)-(+)-4-(3-AMINO-PYRROLIDINO)-7-NITROBENZOFURAZAN BIOREAGENT, SUITABLE FOR FLUORESCENCE, >= 98.0% (HPLC) 1 * 10 mg
Catalog # SIAL76488-10MG-F
Supplier: Merck
(S)-(+)-4-(3-AMINO-PYRROLIDINO)-7-NITROBENZOFURAZAN BIOREAGENT, SUITABLE FOR FLUORESCENCE, >= 98.0% (HPLC) 1 * 10 mg
Catalog # SIAL76488-10MG-F
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection