BIS(2,4,6-TRICHLOROPHENYL) OXALATE BIOREAGENT, SUITABLE FOR CHEMILUMINESCENCE, >= 99.0% (AT) 1 * 5 g
SIAL75707-5G
: Merck
BIS(2,4,6-TRICHLOROPHENYL) OXALATE BIOREAGENT, SUITABLE FOR CHEMILUMINESCENCE, >= 99.0% (AT) 1 * 5 g
SIAL75707-5G
: Merck
Frequently Bought Together
Hydrogen peroxide 35%, J.T.Baker®
: Avantor
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Toluene ≥98%, TECHNICAL
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals