LYSOZYME FROM HEN EGG WHITE LYO.POWDER S ALTF.1000000 U/MG 1 * 5 g
Catalog # SIAL62970-5G-F
Supplier: Merck
LYSOZYME FROM HEN EGG WHITE LYO.POWDER S ALTF.1000000 U/MG 1 * 5 g
Catalog # SIAL62970-5G-F
Supplier: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Optical bottom plates, 96- and 384-well, Nunc™
Supplier: Thermo Fisher Scientific
Eppendorf twin.tec® PCR Plates 384-Well, 45 µl
Supplier: EPPENDORF