Gelest, Bis(triethoxysilylpropyl)amine, 95%, 25g. 1 * 25 g
Catalog # ABCRAB153279.0025
Supplier: abcr
Gelest, Bis(triethoxysilylpropyl)amine, 95%, 25g. 1 * 25 g
Catalog # ABCRAB153279.0025
Supplier: abcr
Frequently Bought Together
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Nitrobenzene 99+% ACS
Supplier: Thermo Fisher Scientific