ContactUsLinkComponent
bacterial pe lb kit has been developed for the extraction of biologically active soluble proteins, including recombinant proteins a nd inclusion bodies, from bacterial cells. bacterial pe lb is a proprietary improvement on the lysozyme based lysis 1 * 500 mL
Catalog # 786-191
Supplier:  G-Biosciences
bacterial pe lb kit has been developed for the extraction of biologically active soluble proteins, including recombinant proteins a nd inclusion bodies, from bacterial cells. bacterial pe lb is a proprietary improvement on the lysozyme based lysis 1 * 500 mL
Catalog # 786-191
Supplier:  G-Biosciences