ContactUsLinkComponent
bacterial pe lb has been developed for the extraction of soluble proteins and inclusion bodies from bacterial cells. it is a propri etary improvement on the lysozyme based lysis, which allows extraction of soluble proteins and concurrent removal of 1 * 500 items
Catalog # 786-188
Supplier:  G-Biosciences
bacterial pe lb has been developed for the extraction of soluble proteins and inclusion bodies from bacterial cells. it is a propri etary improvement on the lysozyme based lysis, which allows extraction of soluble proteins and concurrent removal of 1 * 500 items
Catalog # 786-188
Supplier:  G-Biosciences