ContactUsLinkComponent
bacterial pe lb has been developed for the extraction of soluble proteins and inclusion bodies from bacterial cells. it is a propri etary improvement on the lysozyme based lysis, which allows extraction of soluble proteins and concurrent removal of 1 * 100 mL
Catalog # 786-185
Supplier:  G-Biosciences
bacterial pe lb has been developed for the extraction of soluble proteins and inclusion bodies from bacterial cells. it is a propri etary improvement on the lysozyme based lysis, which allows extraction of soluble proteins and concurrent removal of 1 * 100 mL
Catalog # 786-185
Supplier:  G-Biosciences