ContactUsLinkComponent
Detergent ASB-C8 (4-n-Octylbenzoylamido-propyl-dimethylammonio sulfobetaine). An aminosulfobetaine zwitterionic detergent with a C8 alkyl tail. 1 * 5 g
Catalog # DG065
Supplier:  G-Biosciences
Detergent ASB-C8 (4-n-Octylbenzoylamido-propyl-dimethylammonio sulfobetaine). An aminosulfobetaine zwitterionic detergent with a C8 alkyl tail. 1 * 5 g
Catalog # DG065
Supplier:  G-Biosciences
Frequently Bought Together