ContactUsLinkComponent
immobilized boronic acid is designed for the isolation ofribonucleotide and oligonucleotide rna. the resin consists of boronic acid covalently linked to a polyacrylamide support that offers simple isolation of small molecular weight compounds that ha 1 * 10 mL
Catalog # 786-313
Supplier:  G-Biosciences
immobilized boronic acid is designed for the isolation ofribonucleotide and oligonucleotide rna. the resin consists of boronic acid covalently linked to a polyacrylamide support that offers simple isolation of small molecular weight compounds that ha 1 * 10 mL
Catalog # 786-313
Supplier:  G-Biosciences