ContactUsLinkComponent
Beaker, volume 600ml, material quartzglass, diameter 70mm, height 145mm, tall form, with spout, as per DIN 12330. 1 * 1 items
Catalog # QUTW1231
Beaker, volume 600ml, material quartzglass, diameter 70mm, height 145mm, tall form, with spout, as per DIN 12330. 1 * 1 items
Catalog # QUTW1231
Frequently Bought Together