ContactUsLinkComponent
Areometer, DIN12791, M50 series, measuring range: 1.75-1.80:0.001g/cm , without thermometer, reference temperature:20°C, length:335m m, suitable for government verification, with works certificate. 1 * 1 items
Catalog # AMARH808006/WC
Supplier:  Amarell
Areometer, DIN12791, M50 series, measuring range: 1.75-1.80:0.001g/cm , without thermometer, reference temperature:20°C, length:335m m, suitable for government verification, with works certificate. 1 * 1 items
Catalog # AMARH808006/WC
Supplier:  Amarell
Frequently Bought Together