ContactUsLinkComponent
Rack, XY-rack, material PP, length 206mm, width 139mm, height 27mm, blue, diameter 12mm, temperature resistant -80°C, used on both s ides rack with 96 places for PCR or 0.5ml tube on one side and 1.5/2ml tubes on the other side. 1 * 1 items
Catalog # KOEH2-1960
Rack, XY-rack, material PP, length 206mm, width 139mm, height 27mm, blue, diameter 12mm, temperature resistant -80°C, used on both s ides rack with 96 places for PCR or 0.5ml tube on one side and 1.5/2ml tubes on the other side. 1 * 1 items
Catalog # KOEH2-1960
Frequently Bought Together