Pyrrole ≥98%
Glass bottle for solids, 50 g
Dodavatel: Thermo Fisher Scientific
Přejít na:
Product Details & Documents
Purity: 98+% CAS No.: 109-97-7 Molecular Formula: C4H5N Formula Weight: 67.09 Storage Temperature: 2°C to 8°C Flash Point: 38°(100°F) Refractive Index: 1.5090 Odour: Mild Boiling Point: 131° Beilstein Reference: 1159 Merck Reference: 14,8014 UN No.: 1992 MDL No.: MFCD00005216
Synonyma: 1H-Pyrrole
- Číslo CAS:109-97-7
- Vzorec:C₄H₅N
- MDL číslo:MFCD00005216
- Molecular Weight:67.09 g/mol
- Merck Index:14,8014
- Bod varu:131 °C
- Bod tání:–24 °C
- Hustota:0.967
- Bod vzplanutí:38 °C (100 °F)
- EINECS:203-724-7
- UN:1992
- ADR:3,III
Documents
Specifikace
Specifikace
- Pack type:Glass bottle for solids
- Pk:50 g
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



