METHYL TRIDECANOATE 1 * 5 g
Catalog # SIALT0627-5G
Supplier: Merck
Frequently Bought Together
Aniline for synthesis, Sigma-Aldrich®
Supplier: Merck
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 100…120 °C, AnalaR NORMAPUR® ACS analytical reagent
Supplier: VWR Chemicals
Diethanolamine, EMSURE® for analysis, Supelco®
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
N-(1-Naphthyl)ethylenediamine dihydrochloride, Supelco®
Supplier: Merck
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Iodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
2-Ethoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
Acetone ≥99%
Supplier: Thermo Fisher Scientific
n-Heptane 95%
Supplier: Thermo Fisher Scientific