ContactUsLinkComponent
Pipette, 2:0.02ml, graduated, class AS, DIN 12697 ISO product with single certificate 1 * 1 items
Catalog # HIRS110014108
Supplier:  HIRSCHMANN
Pipette, 2:0.02ml, graduated, class AS, DIN 12697 ISO product with single certificate 1 * 1 items
Catalog # HIRS110014108
Supplier:  HIRSCHMANN
Frequently Bought Together