Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
2,6-Dibromopyridine ≥98%

2,6-Dibromopyridine ≥98%

Glass bottle for solids, 25 g
Catalog # A15397.14
Toxic
Irritant
2,6-Dibromopyridine ≥98%
Catalog # A15397.14
CAS Number:  626-05-1
2,6-Dibromopyridine ≥98%
2,6-Dibromopyridine ≥98%

Glass bottle for solids, 25 g

CAS Number:  626-05-1
Price
1 760,00 Kč
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 626-05-1 Molecular Formula: C5H3Br2N Formula Weight: 236.90 Flash Point: 213°(415°F) Odour: Recognizable Boiling Point: 255° Beilstein Reference: 108922 UN No.: 2811 MDL No.: MFCD00006223

  • CAS Number:626-05-1
  • Formula:C₅H₃Br₂N
  • MDL Number:MFCD00006223
  • Molecular Weight:236.89 g/mol
  • Boiling Pt:255 °C
  • Melting Pt:117…120 °C
  • Flash Pt:213 °C (415 °F)
  • Storage Temperature:Ambient
  • EINECS:210-926-9
  • UN:2811
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.