0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Flask volumetric, capacity 10ml, USP individual certificate, class A, graduated, conformity certificate. 1 * 1 items
HIRS282016145
:
HIRSCHMANN
Flask volumetric, capacity 10ml, USP individual certificate, class A, graduated, conformity certificate. 1 * 1 items
HIRS282016145
:
HIRSCHMANN
Frequently Bought Together
VWR® Volumetric Flasks, Class A, Glass, Clear, NS Plastic Stopper
: VWR Collection
VWR®, Bulb Pipettes, One Mark, Class AS
: VWR Collection
VWR®, Hydrometers, M100
: VWR Collection
VWR®, Measuring Flasks With Single Certificate
: VWR Collection
Graduated pipettes, with piston
: HIRSCHMANN
VWR® Measuring Cylinders, Low Form, Class B, Borosilicate Glass
: VWR Collection
VWR® Volumetric Flasks, Class A, Glass, Amber, NS Plastic Stopper
: VWR Collection
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
: VWR Collection
Measuring cylinders, tall form, class A, borosilicate glass, USP, BLAUBRAND®
: Brand
VWR® Graduated Pipettes, Type 3, Class AS
: VWR Collection
Flask, volumetric, capacity 25ml, class A, USP certificate 1 * 1 items
: HIRSCHMANN