ContactUsLinkComponent
Flask volumetric, capacity 1l, class A, amber graduated, polystoppered, tolerance 0.4ml, height 300mm, NS-ground 24/29, graduation c olor white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264019308
Supplier:  HIRSCHMANN
Flask volumetric, capacity 1l, class A, amber graduated, polystoppered, tolerance 0.4ml, height 300mm, NS-ground 24/29, graduation c olor white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264019308
Supplier:  HIRSCHMANN
Frequently Bought Together