ContactUsLinkComponent
Flask volumetric, capacity 500ml, class A, amber graduated, polystoppered, tolerance 0.25ml, height 260mm, NS-ground 19/26, graduati on color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264019008
Supplier:  HIRSCHMANN
Flask volumetric, capacity 500ml, class A, amber graduated, polystoppered, tolerance 0.25ml, height 260mm, NS-ground 19/26, graduati on color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264019008
Supplier:  HIRSCHMANN
Frequently Bought Together