ContactUsLinkComponent
Flask volumetric, capacity 250ml, class A, amber graduated, polystoppered, tolerance 0.15ml, height 220mm, NS-ground 14/23, graduati on color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264018608
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 250ml, class A, amber graduated, polystoppered, tolerance 0.15ml, height 220mm, NS-ground 14/23, graduati on color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264018608
Supplier:  HIRSCHMANN
Frequently Bought Together