ContactUsLinkComponent
Flask volumetric, capacity 50ml, class A, amber graduated, polystoppered, tolerance 0.08ml, height 140mm, NS-ground 14/23, draduatio n color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264017808
Supplier:  HIRSCHMANN
Flask volumetric, capacity 50ml, class A, amber graduated, polystoppered, tolerance 0.08ml, height 140mm, NS-ground 14/23, draduatio n color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264017808
Supplier:  HIRSCHMANN
Frequently Bought Together