COUPLING FOR LID WITH O-RING RED 1 * 1 items
Catalog # INTG158424
Dodavatel: Integra
Frequently Bought Together
Schutzfilter 5 ml für Eppendorf Research Plus 1 * 50 ST
Dodavatel: EPPENDORF
Acetone ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
VWR® Standard Line Transfer Pipettes
Dodavatel: VWR Collection
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
VWR®, Accessories for Mechanical Pipettes
Dodavatel: VWR Collection
VWR®, Digital Timer, Four Channels, with Clock
Dodavatel: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR® Premium Line, Serological Pipettes
Dodavatel: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
VWR® Serological Pipette, Standard Line
Dodavatel: VWR Collection
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Dodavatel: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Dodavatel: VWR Collection
COUPLING FOR LID WITH O-RING BLUE 1 * 1 items
Dodavatel: Integra
Vacuum seal PTFE / FKM
Dodavatel: Heidolph Instruments GmbH & Co.KG