Rutin hydrate certified 1 * 0,5 g
Catalog # EHERC16880000
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
Buffer solutions
Supplier: Hanna
VWR® Buffer Solutions, Secondary Standards
Supplier: VWR Chemicals
Sulphuric acid 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
Buffer solutions
Supplier: REAGECON