PLATINUM(0)-1,3-DIVINYL-1,1,3,3-TETRAMEDI-SILOXANE COMPLEX, SOLUTION IN XYLENES (PT2%) 1 * 5 g
Catalog # SIAL479519-5G
Supplier: Merck
PLATINUM(0)-1,3-DIVINYL-1,1,3,3-TETRAMEDI-SILOXANE COMPLEX, SOLUTION IN XYLENES (PT2%) 1 * 5 g
Catalog # SIAL479519-5G
Supplier: Merck
Frequently Bought Together
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
2-Methylpentane, Sigma-Aldrich®
Supplier: Merck
Isobutyronitrile 99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
2-Butoxyethanol ≥99%
Supplier: Thermo Fisher Scientific
1-Pentanol 99%, pure
Supplier: Thermo Fisher Scientific
Ethylene glycol, high purity
Supplier: VWR Chemicals
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
1-Butanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Diethyl ether stabilised, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
1-Pentanol 99+% ACS
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals