Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Vials with screw thread, Nalgene®

Vials with screw thread, Nalgene®

Serum vial, 5,00 ml, Clear, No, 20-415, +
Catalog # 215-1497
   Low Manufacturing Impact
Vials with screw thread, Nalgene®
Catalog # 215-1497
   Low Manufacturing Impact
Vials with screw thread, Nalgene®
Vials with screw thread, Nalgene®

Serum vial, 5,00 ml, Clear, No, 20-415, +

Přejít na:
Price
34 650,00 Kč
Přihlásit se pro dostupnost
1 * 1.656 KS
Product Details & Documents

These vials are ideal for diagnostic and biological sera, pharmaceuticals for veterinary and ophthalmologic usage as well as many other applications. They are made from clear or coloured brown PETG.

  • Available clear or coloured brown, sterile or non-sterile
  • A wide range of different screw caps is optionally available
  • Manufactured and packed in workrooms equipped with HEPA filters

Closures for vials: HDPE, coloured. For safe and tight closure of narrow-neck vials. Available in non-sterile or sterile versions. Non sterile caps are loose packed, sterile caps are loose packed in a box with a three-section insert.

For safe and tight closure of narrow-neck vials.

Documents

Specifikace
  • Cap included:
    No
  • Cap Size:
    20-415
  • Sterile:
    +
  • Capacity:
    5,00 ml
  • Colour:
    Clear
  • Description:
    Serum vial
  • Cat. No.:
    215-1497
  • Environmental certification:
    NAR Certificate of Retirement, ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact