ContactUsLinkComponent
Test Sieve 400/65 mm, mesh = 0.071 mm ISO 3310-1 1 * 1 items
  FRIT34.5800.02
 :  FRITSCH
Test Sieve 400/65 mm, mesh = 0.071 mm ISO 3310-1 1 * 1 items
  FRIT34.5800.02
 :  FRITSCH
Frequently Bought Together