ContactUsLinkComponent
Laboratory conductivity meter Lab 970 Set. Sensor recognition. Measuring ranges 0.000 µS/cm...500 mS/cm, salinity, total dissolved s olids (TDS), temp., RS 232 C and USB (slave) interface, micropr., GLP conform. Incl. cover Z 880, stand S4D Z 865, po 1 * 1 items
Catalog # 663-0108
Supplier:  SI Analytics
Laboratory conductivity meter Lab 970 Set. Sensor recognition. Measuring ranges 0.000 µS/cm...500 mS/cm, salinity, total dissolved s olids (TDS), temp., RS 232 C and USB (slave) interface, micropr., GLP conform. Incl. cover Z 880, stand S4D Z 865, po 1 * 1 items
Catalog # 663-0108
Supplier:  SI Analytics
Frequently Bought Together