ContactUsLinkComponent
Areometer, BS718, L50 series SP precision, measuring range:0.700-0.750:0.0005g/ml, without thermometer, length:335mm, reference temp erature:15°C, suitable for government verification. 1 * 1 items
Catalog # AMARH806304
Supplier:  Amarell
Areometer, BS718, L50 series SP precision, measuring range:0.700-0.750:0.0005g/ml, without thermometer, length:335mm, reference temp erature:15°C, suitable for government verification. 1 * 1 items
Catalog # AMARH806304
Supplier:  Amarell
Frequently Bought Together