Haloxyfop-R-methyl certified 1 * 0,1 g
Catalog # EHERC14062500
Supplier: EHRENSTORFER
Frequently Bought Together
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
UV/Visible standards, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach