ContactUsLinkComponent
SUCTION TUBE WITH SIDE HOSE CONNECTION LENGTH 100MM, GL14, SCREW CAP WITH HOLE, GASKET W.PTFE-WASHER12X6MM 1 * 1 items
Katalogové číslo WITG2.385.014
Supplier:  witeg Labortechnik
SUCTION TUBE WITH SIDE HOSE CONNECTION LENGTH 100MM, GL14, SCREW CAP WITH HOLE, GASKET W.PTFE-WASHER12X6MM 1 * 1 items
Katalogové číslo WITG2.385.014
Supplier:  witeg Labortechnik
Frequently Bought Together