SCREW CAP PHENOLIC 15MM BUTYL/PTFE 1 * 100 items
Catalog # SCERKSSR15415BY100
Supplier: DWK Life Sciences
SCREW CAP PHENOLIC 15MM BUTYL/PTFE 1 * 100 items
Catalog # SCERKSSR15415BY100
Supplier: DWK Life Sciences
Frequently Bought Together
Barrier tips, extended, ART™
Supplier: Thermo Fisher Scientific
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Deep Well Plates, Square Well
Supplier: VWR Collection
Test tube, no rim, sodaglass, 40x9/10x0.7mm. 1 * 1.000 items
Supplier: DWK Life Sciences
Containers, wide mouth, with screw cap, PACKO
Supplier: CURTEC
Eppendorf® Protein LoBind Plates
Supplier: EPPENDORF
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Jars, wide mouth, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Separate Snap Caps
Supplier: VWR Collection