ContactUsLinkComponent
FLASK,ROUND BOTTOM,50ML,3 NECKS,BOROSIICATE GLASS,CENTRE NECK ST19/26,TWO ANGLED SIDE NECKS 20° ST14/23 1 * 1 items
Catalog # WITG0.727.050
Supplier:  witeg Labortechnik
FLASK,ROUND BOTTOM,50ML,3 NECKS,BOROSIICATE GLASS,CENTRE NECK ST19/26,TWO ANGLED SIDE NECKS 20° ST14/23 1 * 1 items
Catalog # WITG0.727.050
Supplier:  witeg Labortechnik
Frequently Bought Together