Model 3510 pH meter supplied with glass combination electrode (924 005), electrode holder, ATC probe (027 500), pH 4, 7 and 10 buffer sachets, 230V leaded power supply, BNC shorting plug (009 146), condensed operating instructions and instruction man 1 * 1 items
Catalog # JENW351301
Supplier: Cole-Parmer
Frequently Bought Together