ContactUsLinkComponent
Thermometer, digital, electronic, measuring range:-50:200/0.1C, piercing probe of stainless steel 300x3.5mm, battery life:500hrs, d imension: 22x55mm, hold-function, automatic power-off after 10 min. 1 * 1 items
Catalog # AMARE905004
Supplier:  Amarell
Thermometer, digital, electronic, measuring range:-50:200/0.1C, piercing probe of stainless steel 300x3.5mm, battery life:500hrs, d imension: 22x55mm, hold-function, automatic power-off after 10 min. 1 * 1 items
Catalog # AMARE905004
Supplier:  Amarell
Frequently Bought Together