ContactUsLinkComponent
Jetpei-fluof dna transfection reagent is a fluorescein-conjugated linear polyethylenimine derivative (excitation at 490 nm emission at 520 nm) 1 * 500 µl
Catalog # 105-05N
undefined
Jetpei-fluof dna transfection reagent is a fluorescein-conjugated linear polyethylenimine derivative (excitation at 490 nm emission at 520 nm) 1 * 500 µl
Catalog # 105-05N