Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Filter tubes, plain neck

Filter tubes, plain neck

18 mm, 150 mm
Catalog # BIBB2110/06M
Supplier:   SciLabware
Filter tubes, plain neck
Catalog # BIBB2110/06M
Supplier:  SciLabware
Filter tubes, plain neck
Filter tubes, plain neck

18 mm, 150 mm

Price
20 800,00 Kč
Log in for availability
1 * 50 KS
Product Details & Documents

These filter tubes can be used for small scale filtrations. They are ideal for use with Pyrex® short stem funnels.

Specifications
  • Tube ext.Ø:
    18 mm
  • Seq:
    01
  • Tube length:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.